CLS3081150LO
Pyrex® small knob top desiccator
Lower only
Sinónimos:
equipment replacement accessory
About This Item
Código UNSPSC:
41104510
NACRES:
NB.43
Productos recomendados
Materiales
borosilicate glass
fabricante / nombre comercial
Corning 3081-150LO
Categorías relacionadas
Descripción general
This 2.4 L Pyrex desiccator is designed for general analytical work. The sturdy knob makes the cover easy to handle.
Warning: Do not heat or subject to pressure. Not for vacuum applications.
Warning: Do not heat or subject to pressure. Not for vacuum applications.
This is the desiccator bottom replacement which fits Pyrex desiccator CLS3181150.
Información legal
PYREX is a registered trademark of Corning, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)