Merck
38956
Supelco
Simvastatin
analytical standard
About This Item
Fórmula empírica (notación de Hill):
C25H38O5
Número de CAS:
Peso molecular:
418.57
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
grade
analytical standard
Quality Level
assay
≥98.0% (HPLC)
optical activity
[α]/D +275±25°, c = 1 in acetonitrile
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
impurities
≤0.5% water
mp
127-132 °C (lit.)
application(s)
forensics and toxicology
pharmaceutical (small molecule)
veterinary
format
neat
storage temp.
2-8°C
SMILES string
[H][C@]12[C@H](C[C@@H](C)C=C1C=C[C@H](C)[C@@H]2CC[C@@H]3C[C@@H](O)CC(=O)O3)OC(=O)C(C)(C)CC
InChI
1S/C25H38O5/c1-6-25(4,5)24(28)30-21-12-15(2)11-17-8-7-16(3)20(23(17)21)10-9-19-13-18(26)14-22(27)29-19/h7-8,11,15-16,18-21,23,26H,6,9-10,12-14H2,1-5H3/t15-,16-,18+,19+,20-,21-,23-/m0/s1
InChI key
RYMZZMVNJRMUDD-HGQWONQESA-N
Gene Information
human ... HMGCR(3156)
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Biochem/physiol Actions
Simvastatin is a specific inhibitor of HMG-CoA reductase, the enzyme that catalyzes the conversion of HMG-CoA to mevalonate, an early step in cholesterol biosynthesis. It is used in the treatment of hypercholesterolemia, as it reduces levels of low-density lipoproteins and triglycerides, and raises high-density lipoprotein levels. Simvastatin is a lactone that is readily hydrolyzed in vivo to the corresponding β-hydroxyacid, and can be activated prior to use with NaOH in EtOH treatment. It is a synthetic analog of lovastatin (Cat. No. M2147).
pictograms
Health hazard
signalword
Warning
hcodes
Hazard Classifications
Repr. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Jennifer G Robinson et al.
JAMA, 311(18), 1870-1882 (2014-05-16)
In phase 2 studies, evolocumab, a fully human monoclonal antibody to PCSK9, reduced LDL-C levels in patients receiving statin therapy. To evaluate the efficacy and tolerability of evolocumab when used in combination with a moderate- vs high-intensity statin. Phase 3
Laura Marin et al.
Expert opinion on drug delivery, 8(9), 1205-1220 (2011-05-28)
Chronic Obstructive Pulmonary Disease (COPD) is a severe disease that leads to a non-reversible obstruction of the small airways. The prevalence of this disease is rapidly increasing in developed countries, and in 2020 it has been predicted that this disease
L B Ramsey et al.
Clinical pharmacology and therapeutics, 96(4), 423-428 (2014-06-12)
Simvastatin is among the most commonly used prescription medications for cholesterol reduction. A single coding single-nucleotide polymorphism, rs4149056T>C, in SLCO1B1 increases systemic exposure to simvastatin and the risk of muscle toxicity. We summarize evidence from the literature supporting this association
A Marot et al.
Acta clinica Belgica, 66(2), 134-136 (2011-06-03)
Myopathy, including rhabdomyolysis, is a well-known, albeit rare complication of statin therapy. Predisposing factors include comorbidities and the concomitant use of cytochrome P-450 (CYP) 3A4 inhibitors. We report a case of severe simvastatin-induced rhabdomyolysis triggered by the addition of amiodarone
Joost Besseling et al.
JAMA, 313(10), 1029-1036 (2015-03-11)
Familial hypercholesterolemia is characterized by impaired uptake of cholesterol in peripheral tissues, including the liver and the pancreas. In contrast, statins increase the cellular cholesterol uptake and are associated with increased risk for type 2 diabetes mellitus. We hypothesize that