Documentos clave
16811
Supelco
Reference Electrode
for ion-selective electrodes
About This Item
UNSPSC Code:
26111700
NACRES:
NB.61
grade
for ion-selective electrodes
description
S7 connector
General description
Visit our Sensor Applications portal to learn more.
reference electrode with bridge, PTFE diaphragm, filled with 3 M KCl
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)