Merck
MIX0500L101
Millipore
Mobius® MIX500, 230 V, 50/60 Hz
About This Item
UNSPSC Code:
41103801
eCl@ss:
32010600
Components
Includes carrier, control box unit, motor, hand-held control and cable
Legal Information
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany
For use with
Referencia del producto
Descripción
Precios
Visite la Librería de documentos
Artículos
This page provides an overview of USP <87>, <88>, and <1031>, compares bioreactivity testing, and describes the transition of our organization towards in vitro test methods.
This page provides an overview of USP <87>, <88>, and <1031>, compares bioreactivity testing, and describes the transition of our organization towards in vitro test methods.
This page provides an overview of USP <87>, <88>, and <1031>, compares bioreactivity testing, and describes the transition of our organization towards in vitro test methods.
This page provides an overview of USP <87>, <88>, and <1031>, compares bioreactivity testing, and describes the transition of our organization towards in vitro test methods.