Documentos clave
Z677884
Finntip® Flex pipette tips
Finntip Flex 200, non-sterile
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
maximum volume:
200 μL
volume range:
1-200 μL
sterility:
non-sterile
feature:
barrier: no
packaging:
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Finn 94060310
material
colorless tip
yellow
sterility
non-sterile
feature
barrier: no
packaging
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Finn 94060310
maximum volume
200 μL
volume range
1-200 μL
General description
Finntip Flex pipette tips are easy-to-use, soft, flexible high-end tips. They are molded with diamond polished tooling for an extra smooth tip surface, which prevents liquid retention and achieves better pipetting accuracy.
Other Notes
Browse all pipette tips
Legal Information
Finntip is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)