Key Documents
Z549517
Sigma-Aldrich
Stopcock Adapter
straight PTFE stopcock, joint: ST/NS 14/20
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
material
straight PTFE stopcock
joint
joint: ST/NS14/20
General description
Glass and PTFE stopcocks have a 2 mm bore and accommodate 1/4 in. I.D. tubing. Can be used in vacuum or atmospheric applications.