Key Documents
Z530948
Sigma-Aldrich
Aldrich® Liebig condenser
Overall H 60 mm, joint: ST/NS 14/20
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
joint
joint: ST/NS14/20
Overall H
60 mm
General description
Condensers have a standard taper female joint and a lower male drip tip-joint.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC