Key Documents
Z513342
Sigma-Aldrich
Micro spatulas
flat rounded ends, L 8 in. (203 mm)
About This Item
UNSPSC Code:
41122403
NACRES:
NB.35
material
nickel-stainless steel
packaging
pkg of 12 ea
L
8 in. (203 mm)
General description
Nickel stainless spatula with both ends rounded, each end approximately 5/16 in. (8 mm) wide x 2 in. (51 mm) long.
Visite la Librería de documentos
Los clientes también vieron
Slide 1 of 4
1 of 4