Key Documents
Z506850
Sigma-Aldrich
Quick Press SS collar
diam. 7 mm
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
technique(s)
IR spectroscopy: suitable
diam.
7 mm
General description
Replacement part for Z506796 Quick Press die set, diam. 7 mm