Merck
Z283800
®Pulgas agitadoras magnéticas Spinbar
white, diam. 2 mm, L 2 mm
About This Item
Código UNSPSC:
41103806
NACRES:
NB.72
Materiales
PTFE-coated
cylindrical PTFE-coated
Características
ALNICO V coated magnet
envase
pack of 3 ea
fabricante / nombre comercial
Bel-Art F37119-0002
Longitud
2 mm
Diámetro
2 mm
color
white
Descripción general
  • Spinbar® magnetic stirring fleas are designed for use in tubes, vials, and gradient markers.
  • Ideal for mixing of small volume samples in environmental testing and life science applications.
  • Alnico magnet encapsulated in durable and inert FDA grade PTFE (polytetrafluoroethylene) coating.
  • Broad chemical resistance for use in a wide variety of applications.
  • Autoclavable.
Very small, efficient stirring bars. Ideal for vials, micro test tubes, centrifuge tube mixing etc.
  • PTFE-coated
  • ALNICO V coated magnet
Información legal
Spinbar is a registered trademark of SP Scienceware, a division of SP
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)