Documentos clave
Z281417
Sigma-Aldrich
Uvex® Stealth safety goggles
teal frame
About This Item
UNSPSC Code:
46181802
NACRES:
NB.35
availability
not available in EU
not available in EU
color
teal frame
General description
  • Wide curve antifog lenses for unrestricted field of view
  • Unique ventilation system keeps out splashes, particulates, and vapors
  • Internal silicone rubber cushion provides full-day comfort
Clear PC lens with 4-C coating (antifog, antistatic, antiscratch, and UV). UV protection blocks 99.9% of harmful UV rays. Meets ANSI Z87.1-1989 and exceeds ANSI Z80.3 requirements.
Legal Information
Stealth is a trademark of Bacou-Dalloz Eye & Face Protection, Inc.
Uvex is a registered trademark of Uvex Winter Optik GmbH Ltd.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)