Key Documents
Z278378
Sigma-Aldrich
Scienceware® gas-washing bottle
for tubing with a 1/4 in. O.D.
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
vapor pressure
≤0.5 psi
feature
for tubing with a 1/4 in. O.D.
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H11030-0000
General description
An economical device for precipitating materials from gas. Graduated 250 mL PP cylinder with 70 μm porosity dispersion disk. Vinyl stopper has inlet and outlet ports for gas.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP