Z233463
Aldrich® Schlenk-type adapter
Offset with septum-inlet, male joint: ST/NS 14/20
About This Item
Código UNSPSC:
41121815
NACRES:
NB.43
Productos recomendados
Junta
female joint ST/NS14/20
male joint: ST/NS14/20
Descripción general
Thermoplastic screw caps with Viton O-rings usable to 140 deg C.
Ligadura / enlace
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Información legal
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)