Key Documents
Z222038
Bransonic® ultrasonic cleaner accessories
Perforated Tray, stainless steel, tank capacity 2.8 L
About This Item
UNSPSC Code:
41101500
NACRES:
NB.74
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Branson 100-410-162
tank capacity
2.8 L
General description
Perforated stainless steel tray for use with Branson M series, model 2800 Utrasonic Bath.
Legal Information
Bransonic is a registered trademark of Branson Ultrasonics Corp.