N-Methylpyrrolidone (NMP) is compatible with this product. It does not cause any damage or degradation to the glass surface. It is not recommended to heat the TLC-developing tanks above 100 degrees Celsius. Heating past this temperature may also cause incompatibility issues with NMP.
Z204153
Tanques completos rectangulares de desarrollo de la TLC
L × H × W 17.5 cm × 16.0 cm × 8.2 cm
About This Item
Productos recomendados
técnicas
thin layer chromatography (TLC): suitable
L × Al × An
17.5 cm × 16.0 cm × 8.2 cm
Nota al pie de página
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Los clientes también vieron
-
Is this NMP compatible? And can it be heated?
1 answer-
Helpful?
-
-
How thick are the walls of product Z204153?
1 answer-
Glass thickness is 0.34 to 0.20 mm.