The 5ml Pipette Bulb (product Z186678) is a natural rubber bulb with a capacity of 5ml. It has the following dimensions:Bore Size: 4mmDiameter: 29mmLength: 41mm
Z186678
Pipette bulb
capacity 5 mL
About This Item
Productos recomendados
Materiales
natural rubber
round red
envase
pkg of 3 ea
fabricante / nombre comercial
200-5
capacidad
5 mL
Categorías relacionadas
Descripción general
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Los clientes también vieron
-
What are the dimensions for the 5ml Pipette Bulb (product Z186678)?
1 answer-
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.