Key Documents
Z123544
Sigma-Aldrich
Econo-Cards
About This Item
UNSPSC Code:
12000000
technique(s)
IR spectroscopy: suitable
General description
13mm KBr pellets or paper inserts mount into card for identification and storage.