Z115533
Scienceware® protective lab-labeling system labels
size 1 1/2 in. × 3 in.
About This Item
Código UNSPSC:
55121611
NACRES:
NB.35
Productos recomendados
Materiales
paper
white
envase
pkg of 10 (100 labels per pad)
fabricante / nombre comercial
Bel-Art F13455-0015
tamaño
1 1/2 in. × 3 in.
color
white
Descripción general
Adhesive is resistant to casual exposure to acids, alkalis, and organic solvents. Temp range: -196 to 120 °C.
For a waterproof covering, purchase protective lab-labeling system tape to protect these labels.
For a waterproof covering, purchase protective lab-labeling system tape to protect these labels.
Información legal
Scienceware is a registered trademark of SP Scienceware, a division of SP
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)