Z101311
Aldrich® septum-inlet adapter with side-arm
joint: ST/NS 19/22
About This Item
Productos recomendados
joint
joint: ST/NS19/22
General description
Septum-Einlässe benötigen Septum Z10,072-2 oder Z12,435-4 sofern nicht mit * gekennzeichnet.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC