Key Documents
TEM-CF150-CU-TH
Sigma-Aldrich
Carbon Film Supported Copper Square Mesh
Thick, size 150 mesh, box of 50
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 50
size
150 mesh
General description
A 10 nm thick film of pure carbon supported standard squre mesh grids. Copper grid, 150 mesh.