Documentos clave
SYNC151110
Synthware West condenser
joint: ST/NS 14/20, jacket L 110 mm
Sinónimos:
Synthware C151110, glass West condenser
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C151110
jacket L
110 mm
General description
With narrow annular space for fast coolant speed, thus high cooling efficiency.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)