Key Documents
HAM59000-45
Hamilton® Microlab® 600 instrument syringe
model, 1005 BFP, volume 5 mL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
Gastight Borosilicate glass/PTFE syringe
feature
model 1005 BFP
manufacturer/tradename
Hamilton 59000-45
parameter
10-115 °C temp. range (50-239 °F)
200 psig pressure
I.D. × O.D.
10.3 mm × 13.5 mm
volume
5 mL
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Microlab is a registered trademark of Hamilton Co.