CAD9926
Animal feeding needle, sterile, disposable, PTFE
size 17G, L × diam. 3.3 in. × 2.4 mm , ball
About This Item
Código UNSPSC:
42142609
NACRES:
NB.03
Productos recomendados
tamaño
17G
envase
pkg of 20 x 5 ea (5 needles per sterile Tyvek peel pouch - 20 pouches per box)
L × diám.
3.3 in. × 2.4 mm , ball
Descripción general
- PTFE tube is softer than other tubing types, very flexible, and translucent for easy confirmation of reagent delivery
- Plastic Luer hubs and soft silicone tips
- Non-toxic and no-pyrogenic
- Packaged 5 needles per sterile Tyvek peel pouch - 20 pouches per box
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)