BR65038
BRAND® desiccator ′Novus′
nominal diameter × O.D. × H 200 mm × 270 mm × 298 mm, with knob lid
About This Item
UNSPSC Code:
41104510
NACRES:
NB.43
Productos recomendados
feature
with knob lid
manufacturer/tradename
BRAND 65038
nominal diameter × O.D. × H
200 mm × 270 mm × 298 mm
Categorías relacionadas
General description
Boro 3.3. DIN 12491. Precisely ground flat flanges. Base ring surface studded to improve stability and help prevent cracks. Interchangeable lid.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG