BAF249660006
SP Bel-Art® Splash Shield Table Mounting Fixture
fixed table bracket, attaches permanently with screws to bench top
About This Item
Código UNSPSC:
41122800
NACRES:
NB.43
Productos recomendados
envase
pack of 1 ea
fabricante / nombre comercial
Bel-Art F24966-0006
Descripción general
No matter how your workspace is configured, these Plexiglas® Splash Shields are designed to provide safety plus comfort (not hot and stuffy like personal face shields), flexibility (they’re fully adjustable), and quality in a design that will adapt to your specific needs.
Características y beneficios
- All shields move side-to-side as well as up and down at the touch of a finger
- Free precious work space and enhance the performance of every user
- After selecting desired shield, order appropriate mounting fixtures to suit your work area
Información legal
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Plexiglas is a registered trademark of Arkema
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)