Skip to Content
Merck
All Photos(1)

Documents

MAB8755

Sigma-Aldrich

Anti-Venezuelan Eq. Encephalitis Antibody, clone 1A3B-7

clone 1A3B.7, Chemicon®, from mouse

Synonym(s):

VEE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1A3B.7, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
neutralization: suitable

isotype

IgG2a

shipped in

wet ice

Specificity

Strain PTF-39. Cross reactive across the VEE group.

Immunogen

VEE

Application

Detect Venezuelan Eq. Encephalitis using this Anti-Venezuelan Eq. Encephalitis Antibody, clone 1A3B-7 validated for use in ELISA, IF, IH, NEUT.
ELISA

Indirect Immunohistochemistry

Neutralization

Optimal working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Format: Purified
Purified

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months.

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 6/12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Forrest Keck et al.
Virulence, 9(1), 1403-1421 (2018-08-14)
Venezuelan equine encephalitis virus (VEEV) is a neurotropic arbovirus that is highly infectious as an aerosol and can result in an encephalitic phenotype in infected individuals. VEEV infections are known to be associated with robust inflammation that eventually contributes to
Lauren E Williamson et al.
Cell, 184(17), 4430-4446 (2021-08-21)
Alphaviruses cause severe arthritogenic or encephalitic disease. The E1 structural glycoprotein is highly conserved in these viruses and mediates viral fusion with host cells. However, the role of antibody responses to the E1 protein in immunity is poorly understood. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service