Merck
  • Therapeutic alphavirus cross-reactive E1 human antibodies inhibit viral egress.
Therapeutic alphavirus cross-reactive E1 human antibodies inhibit viral egress.
Cell (2021-08-21)
Lauren E Williamson, Kristen M Reeder, Kevin Bailey, Minh H Tran, Vicky Roy, Mallorie E Fouch, Nurgun Kose, Andrew Trivette, Rachel S Nargi, Emma S Winkler, Arthur S Kim, Christopher Gainza, Jessica Rodriguez, Erica Armstrong, Rachel E Sutton, Joseph Reidy, Robert H Carnahan, W Hayes McDonald, Clara T Schoeder, William B Klimstra, Edgar Davidson, Benjamin J Doranz, Galit Alter, Jens Meiler, Kevin L Schey, Justin G Julander, Michael S Diamond, James E Crowe
ABSTRACT
Alphaviruses cause severe arthritogenic or encephalitic disease. The E1 structural glycoprotein is highly conserved in these viruses and mediates viral fusion with host cells. However, the role of antibody responses to the E1 protein in immunity is poorly understood. We isolated E1-specific human monoclonal antibodies (mAbs) with diverse patterns of recognition for alphaviruses (ranging from Eastern equine encephalitis virus [EEEV]-specific to alphavirus cross-reactive) from survivors of natural EEEV infection. Antibody binding patterns and epitope mapping experiments identified differences in E1 reactivity based on exposure of epitopes on the glycoprotein through pH-dependent mechanisms or presentation on the cell surface prior to virus egress. Therapeutic efficacy in vivo of these mAbs corresponded with potency of virus egress inhibition in vitro and did not require Fc-mediated effector functions for treatment against subcutaneous EEEV challenge. These studies reveal the molecular basis for broad and protective antibody responses to alphavirus E1 proteins.
Sigma-Aldrich
Anti-Eastern Equine Encephalitis Antibody, Virus, clone 1A4B.6, clone 1A4B.6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Venezuelan Eq. Encephalitis Antibody, clone 1A3B-7, clone 1A3B.7, Chemicon®, from mouse