Skip to Content
Merck
All Photos(1)

Key Documents

361555

Sigma-Aldrich

GSK-3 Inhibitor XIII

The GSK-3 Inhibitor XIII, also referenced under CAS 404828-08-6, controls the biological activity of GSK-3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

GSK-3 Inhibitor XIII, (5-Methyl-1H-pyrazol-3-yl)-(2-phenylquinazolin-4-yl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15N5
CAS Number:
Molecular Weight:
301.35
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C18H15N5/c1-12-11-16(23-22-12)20-18-14-9-5-6-10-15(14)19-17(21-18)13-7-3-2-4-8-13/h2-11H,1H3,(H2,19,20,21,22,23)

InChI key

JYCUVOXSZBECAY-UHFFFAOYSA-N

General description

A potent and ATP-competitive inhibitor of GSK-3 (Ki = 24 nM).
An aminopyrazole compound that acts as a potent and ATP-binding site inhibitor of GSK-3 with a Ki of 24 nM.

Biochem/physiol Actions

Cell permeable: no
Primary Target
GSK-3
Product does not compete with ATP.
Reversible: no
Target Ki: 24 nM against GSK-3

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Pierce, A.C., et al. 2005. J. Med. Chem.48, 1278.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service