Skip to Content
Merck
All Photos(1)

Documents

1.93601

Supelco

Potassium iodide

EMPLURA®

Synonym(s):

Potassium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
MDL number:
UNSPSC Code:
12352302

grade

laboratory grade

vapor pressure

1 mmHg ( 745 °C)

product line

EMPLURA®

form

solid

pH

ca.6.9 (at 20 °C, at 50 g/L)

mp

681 °C (lit.)

solubility

water: soluble ca.1,430 g/L at at 25 °C

density

3.13 g/cm3 at at 20 °C (68 °F)

storage temp.

20-25°C

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

1.93601.0251: 250 g, Minimum order quantity: 20
1.93601.0521: 500 g, Minimum order quantity: 20
1.93601.5021: 5 kg, Minimum order quantity: 4
1.93601.9051: 50 kg, Minimum order quantity: 1

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Inorganics and solvents for basic and standard applications manufactured locally.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service