Skip to Content
Merck
All Photos(1)

Key Documents

1.93201

Supelco

Potassium iodide

for analysis EMPARTA® ACS

Synonym(s):

Potassium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
MDL number:
UNSPSC Code:
12352302
grade:
ACS reagent
form:
solid
solubility:
water: soluble ca.1,430 g/L at 25 °C

grade

ACS reagent

vapor pressure

1 mmHg ( 745 °C)

form

solid

pH

ca.6.9 (at 20 °C, at 50 g/L)

mp

681 °C (lit.)

solubility

water: soluble ca.1,430 g/L at 25 °C

density

3.13 g/cm3

storage temp.

20-25°C

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

1.93201.0121: 100 kg, Minimum order quantity: 1
1.93201.0251: 250 g, Minimum order quantity: 1
1.93201.0521: 500 g, Minimum order quantity: 1
1.93201.5021: 5 kg, Minimum order quantity: 1
1.93201.9051: 50 kg, Minimum order quantity: 1

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Inorganics and solvents for basic and standard applications manufactured locally.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service