1.50012
Hibar® RT HPLC Columns for customized packing
L × I.D. 125 mm × 4.6 mm, please select HPLC sorbent for Column filling
Synonym(s):
Hibar HPLC Column
About This Item
Recommended Products
Product Name
Hibar® 125-4,6, suitable for HPLC
parameter
400 bar pressure
Quality Level
technique(s)
HPLC: suitable
column L × I.D.
12.5 cm × 4 mm
separation technique
supercritical fluid (SFC)
storage temp.
no temp limit
Related Categories
General description
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service