1.09962
Barium chloride solution
c(BaCl₂) = 0.05 mol/l (0.1 N), concentrate for 1000 mL titration solution, Titrisol®
About This Item
Recommended Products
Product Name
Barium chloride solution, for 1000 ml, c(BaCl₂) = 0.05 mol/l (0.1 N) Titrisol®
Quality Level
product line
Titrisol®
form
liquid
reaction suitability
reaction type: Acid-base reactions
concentration
0.05 M
technique(s)
titration: suitable
pH
4.3 (20 °C in H2O)
density
1.18 g/cm3 at 20 °C
storage temp.
15-25°C
Looking for similar products? Visit Product Comparison Guide
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service