Key Documents
Z239135
Sigma-Aldrich
White disposable polystyrene pour-boats
W × L × H 116 mm × 190 mm × 25 mm
About This Item
UNSPSC Code:
41100000
NACRES:
NB.71
Pricing and availability is not currently available.
material
oblong
polystyrene
packaging
pack of 250 ea
manufacturer/tradename
Sigma-Aldrich
W × L × H
116 mm × 190 mm × 25 mm
dish L × W
190 mm × 116 mm
General description
Weighing dish designed to facilitate dispensing and pouring. Smooth uniform walls, flat bottom, usable to 200°F (93°C).
Choose from one of the most recent versions:
Certificates of Analysis (COA)