SYNA652440
Synthware™ long stem inlet adapter
joint: ST/NS 24/40
Synonym(s):
Synthware A652440, glass inlet adapter
All Photos(1)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Recommended Products
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A652440
Related Categories
General description
With standard taper top outer and lower inner joints with sealed in lower tube extending below inner joint.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)