SYNA501420
Synthware™ gas inlet adapter
joint: ST/NS 14/20, O.D. × L 8 mm × 153 mm
Synonym(s):
Synthware A501420, gas inlet adapter, extended drip tube
All Photos(1)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Recommended Products
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A501420
O.D. × L
8 mm × 153 mm
General description
Inner joint with a sealed on lower tube extending 6” below the grind, having the same O.D. tubing on both ends of the grind.
Legal Information
Synthware is a trademark of Kemtech America Inc.