Skip to Content
Merck
All Photos(1)

Key Documents

933333

Sigma-Aldrich

SEM Sample Holder

24 Place SEM Holder

Synonym(s):

24-pin SEM mount

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NA.23

Looking for similar products? Visit Product Comparison Guide

General description

EMS techware

Application

This 24-place holder is made from aluminum and offers a stage travel of 100 mm x 50 mm (which is perfect to be used with the Hitachi VP SEM). The holder has a unique identification number so every hole is identifiable. The holder measures 4 7/16" (112.7 mm) x 2½" (63.5 mm) x ⅜"(10 mm) (L x W x H) and has an ⅛" (3.2 mm) mounting pin.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service