Skip to Content
Merck
All Photos(1)

Key Documents

933309

Sigma-Aldrich

SEM Sample Holder

45° Pin Stub Holder, size 3.2 mm , (1/8”) pin

Synonym(s):

Vertical mount

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

size

3.2 mm , (1/8”) pin

Looking for similar products? Visit Product Comparison Guide

General description

EMS techware

Application

Allows for a higher SE signal without having to tilt the specimen stage and it accommodates any standard 12.7 mm (½") pin stubs with 3.2 mm (⅛") pin.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service