Skip to Content
Merck
All Photos(1)

Documents

245038

Sigma-Aldrich

1-Bromoundecane

98%

Synonym(s):

Undecyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10Br
CAS Number:
Molecular Weight:
235.20
Beilstein:
1098390
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

8.2 (vs air)

Assay

98%

form

liquid

refractive index

n20/D 1.457 (lit.)

bp

137-138 °C/18 mmHg (lit.)

mp

−9 °C (lit.)

density

1.054 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCBr

InChI

1S/C11H23Br/c1-2-3-4-5-6-7-8-9-10-11-12/h2-11H2,1H3

InChI key

IKPSIIAXIDAQLG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Bromoundecane (undecyl bromide) was used in the preparation of Grignard reagent by reacting with Mg in THF (tetrahydrofuran).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective total synthesis of (−)-tetrahydrolipstatin using Oppolzer?s sultam directed aldol reaction.
Kumaraswamy G and Markondaiah B.
Tetrahedron Letters, 49(2), 327-330 (2008)
Anna Parus et al.
Ecotoxicology and environmental safety, 208, 111595-111595 (2021-01-06)
Modern agricultural practices are often based on the use of mixtures of specific herbicides to achieve efficient crop protection. The major drawbacks of commercial herbicidal formulations include the necessity to incorporate toxic surfactants and high volatility of active substances. Transformation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service