Skip to Content
Merck
All Photos(1)

Key Documents

339628

Sigma-Aldrich

Dodecylmagnesium bromide solution

1.0 M in diethyl ether

Synonym(s):

1-Dodecylmagnesium bromide, Bromododecylmagnesium, Dodecylmagnesium bromide (6CI), Laurylmagnesium bromide, n-Dodecylmagnesium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11MgBr
CAS Number:
Molecular Weight:
273.54
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in diethyl ether

density

0.843 g/mL at 25 °C

SMILES string

CCCCCCCCCCCC[Mg]Br

InChI

1S/C12H25.BrH.Mg/c1-3-5-7-9-11-12-10-8-6-4-2;;/h1,3-12H2,2H3;1H;/q;;+1/p-1

InChI key

JSDNASSZWGNUBO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 2

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-29.2 °F - closed cup

Flash Point(C)

-34 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service