L8663
Director-Ready™ pc-myc-CMV-2™
About This Item
Code UNSPSC :
12171500
Nomenclature NACRES :
NA.85
Produits recommandés
Température de stockage
−20°C
Niveau de qualité
Description générale
The Director-Ready� pc-myc-CMV2 Expression Vector is de-phosphorylated after Hind III/Bgl II digestion and is ready for use with the Director Universal Cloning System. pc-myc-CMV2 is for transient, cytoplasmic expression of N-terminal c-myc fusion proteins in mammalian cells.
Informations légales
Director-Ready is a trademark of Sigma-Aldrich Co. LLC
pc-myc-CMV-2 is a trademark of Sigma-Aldrich Co. LLC