Z682187
Ace receiving flask adapter
joint: ST/NS 24/40, fits Büchi® models R-200/R-205
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.43
Produits recommandés
Joint
joint: ST/NS24/40
Fabricant/nom de marque
Büchi equivalent, 40616
Catégories apparentées
Description générale
The receiving flask adapter fits condenser assemblies "C", "CR", "A", and "V" for Büchi® evaporator models R-200/205. Adapter clip: not included.
Informations légales
Büchi is a registered trademark of Büchi Labortechnik AG