Principaux documents
Z549894
Sigma-Aldrich
Aldrich® jacketed three-neck flask
full jacket, capacity 1,000 mL, center joint: ST/NS 24/40
Synonyme(s) :
boiling flask, flask round, flask round bottom
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
round bottom
three-neck
three-neck flask
Joint
center joint: ST/NS24/40
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
pack of 1
Fabricant/nom de marque
Sigma-Aldrich
Capacité
1,000 mL
Description générale
Unique design which incorporates a flush-closing PTFE drain valve eliminating unwanted dead space.
Features:
  • Eliminates dead space for complete stirring of flask contents
  • Allows for complete draining without breaking down flask set-up
  • Liquid cooled or heated with liquid or steam
  • 24/40 side joints
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC