Documents
Z153087
Sigma-Aldrich
Aldrich® jacketed round-bottom flasks
capacity 250 mL
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
round bottom flask
three-neck
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
pack of 1
Fabricant/nom de marque
Sigma-Aldrich
Capacité
250 mL
Description générale
24/40 joint, with two hose connections. Multifunctional-can be water cooled or heated with hot water or steam.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC