SYNA542440
Synthware™ vacuum jacketed distillation head
top joint: ST/NS 10/18, bottom joint: ST/NS 24/40, side joint: ST/NS 24/40
Synonyme(s) :
Synthware A542440, glass distilling head
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Produits recommandés
Joint
bottom joint: ST/NS24/40
side joint: ST/NS24/40
top joint: ST/NS10/18
Conditionnement
pack of 1 ea
Fabricant/nom de marque
A542440
Informations légales
Synthware is a trademark of Kemtech America Inc.