Key Documents
HAM59000-15
Hamilton® Microlab® 600 instrument syringe
model, 1705 BFP, volume 50 μL
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
Matériaux
Gastight Borosilicate glass/PTFE syringe
Caractéristiques
model 1705 BFP
Fabricant/nom de marque
Hamilton 59000-15
Paramètres
10-115 °C temp. range (50-239 °F)
1000 psig pressure
D.I. × D.E.
1.03 mm × 7.75 mm
Volume
50 μL
Informations légales
Hamilton is a registered trademark of Hamilton Co.
Microlab is a registered trademark of Hamilton Co.