BR474300
BRAND® transparent plastic staining trough with two lids
PMP, L × W × H 101 mm × 83 mm × 70 mm
Synonyme(s) :
Microscopy, Staining, Staining Accessories
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
MA.01
Produits recommandés
Matériaux
PMP
Fabricant/nom de marque
BRAND 474300
L × l × H
101 mm × 83 mm × 70 mm
Catégories apparentées
Description générale
The BRAND® transparent plastic staining trough is made from clear PMP, providing visibility for monitoring samples during staining processes. It features two lids: one designed to protect against evaporation when the trough is not in use, and another with a handle slot that facilitates easy handling during the staining process. This thoughtful design enhances usability and efficiency in laboratory settings, making it ideal for various staining applications.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG