Principaux documents
BR472200
BRAND® glass staining trough with lid
soda-lime glass
Synonyme(s) :
Microscopy, Staining, Staining Accessories
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.13
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
soda-lime glass
Description
use with tray for glass staining trough; fits 10 slides
Fabricant/nom de marque
BRAND 472200
L × l × H
105 mm × 85 mm × 70 mm
Description générale
The BRAND® Glass Staining Trough with Lid is designed for efficient staining of up to 10 slides, making it an essential tool for laboratory applications. Constructed from durable soda-lime glass, this trough features dimensions of 76 x 26 mm, providing ample space for slide placement. The included lid helps to minimize contamination and evaporation during the staining process. For a complete staining setup, please note that the staining trough, tray, and wire handle must be ordered separately. This versatile and practical design enhances the efficiency of staining procedures in any laboratory setting.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG