Principaux documents
BAF370152000
SP Bel-Art® CultureTemp 37°C Warming Plate
AC/DC input 120 V, US 3-pin plug
Synonyme(s) :
SP BEL-ART CULTURETEMP 37C WARMING PLATE; 120V, TRYPSIN 37°C WARMING PLATE
About This Item
Code UNSPSC :
41102406
Nomenclature NACRES :
NB.11
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F37015-2000
H × W × depth
57 mm (2.25 in.) × 249 mm (9.75 in.) × 254 mm (10 in.)
Description générale
This highly accurate Warming Plate, set at an optimal 37°C (±1°C), is designed to Trypsinize Cells during cell passaging.
Caractéristiques et avantages
  • Avoids numerous entries into the CO2 incubator which alter its atmosphere, injuring ongoing experiments in the incubated cultures
  • Increases cell viability by keeping cells at an optimal temperature during examination of cell cultures outside of the incubator
  • Can be used to maintain the warmth of reagents inside the hood
  • Built in, 15 minute count down timer keeps track of the time that the trypsin has been on the cells
  • Temperature remains uniform across the entire anodized aluminum warming plate; measures 17.8 D x 20.3cmW (7 x 8”)
  • Fits easily in cell culture hoods or on the workbench
  • UV resistant polypropylene housing will not break down under germicidal UV lights
  • Water resistant and safe for cleaning with Ethanol or IPA
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP