Principaux documents
CLS4133
Corning® DeckWorks pipet tips reload starter kits
1 - 300μL pipet tips, graduated, reload starter kit, natural, non-sterile, polypropylene, 96 tips/deck, 10 decks/pack, 960 tips/pack, 1 packcase and 10 empty racks/case
Synonyme(s) :
pipette tip
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.01
Gamme de volumes:
1-300 μL
Stérilité:
non-sterile
Caractéristiques:
barrier: no
disposable
graduated
Tip format: 96 tips/rack
Tip shape: round
Conditionnement:
pack of 960
Fabricant/nom de marque:
Corning 4133
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
natural tip
polypropylene
Description
Starter Kit
Stérilité
non-sterile
Caractéristiques
barrier: no
disposable
graduated
Tip format: 96 tips/rack
Tip shape: round
Conditionnement
pack of 960
Fabricant/nom de marque
Corning 4133
Gamme de volumes
1-300 μL
Description générale
Corning® DeckWorks Reload Starter Kits
DeckWorks starter kits contain one DeckWorks reload system with standard tips and empty hinged racks. 10 empty racks accompany 10 uL, 200 uL and 300 uL volume tips and 8 empty racks accompany 1000 uL volume tips.
This system is designed to allow you to maximize storage space and help reduce the production and waste of plastic materials. Both the hinged racks and reload decks within this system are manufactured with recycled plastic.
The pipet tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes.
Medical grade resins and production excellence allow for full certification as:
  • Nonpyrogenic
  • Human DNA-free
  • RNase-/DNase-free
Informations légales
Corning is a registered trademark of Corning, Inc.
DeckWorks is a trademark of Corning, Inc.