Principaux documents
BAF188700000
SP Bel-Art® Radioimmunoassay Tube Rack
to hold, 216 x 10-13mm Tubes
Synonyme(s) :
polypropylene rack, tube holder
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.43
Caractéristiques
to hold 216 x 10-13mm Tubes
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F18870-0000
H × L × width
3 in. (76 mm) × 13.5 in. (342 mm) × 9.0625 in. (230 mm)
H de la plateforme
2.25 in. (57 mm)
Description générale
These polypropylene racks provide a practical method of pipetting and storing a large number of samples.
Caractéristiques et avantages
  • Easy tracking of last tube filled by using alternate rows and rotating the tubes
  • Useful in clinical labs for storage of samples
  • Steam autoclavable at 121ºC (250ºF)
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP